Multiple film frame shippers
Lieferant: ENTEGRIS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
E4000851010615 E4002661010615 E4002761010615 E40008310167C2 E40008510167C2 E40027610167C2 E40038010167C2 E40026610167C2 E40027610106_U E4000831010615 E40038010267C2 E4003801010615
ENTEE4000851010615EA 0 EUR InStock
ENTEE4000851010615 ENTEE4002661010615 ENTEE4002761010615 ENTEE40008310167C2 ENTEE40008510167C2 ENTEE40027610167C2 ENTEE40038010167C2 ENTEE40026610167C2 ENTEE40027610106_U ENTEE4000831010615 ENTEE40038010267C2 ENTEE4003801010615
Multiple film frame shippers
Wafer-Handling-Zubehör Wafer-Versandbehälter
Multiple film frame shippers provide a safe and effective way to ship and store up to 25 wafers, or singulated die, mounted to film frames. Special applications include shipping 'diced' or thinned GaAs or GaP wafers as well as storage or shipment of wafers mounted to film frame. Replaceable top cover cushion allows for reuse of shipper. Shippers available for 300 mm, 200 mm and 150 mm film frame applications.
  • Snap-in-place replaceable cushion prevents frame movement and minimises particle transfer during shipping
  • Positive snap latch assures secure closure
  • Non-obtrusive latch and stackable design reduce storage space and shipping costs
  • Rounded corners and sloped sides minimise water entrapment during cleaning
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge