4 TRAY 10x6 ARRAY 2,03 DP H44-260465-66C02 Material: ChipSentry (Black Conductive Polycarbonate) Pocket Dimensions: 0.260 x 0.465 x 0.080 Capacity: 10x6 (60) 1 * 200 ST
Lieferant: ENTEGRIS
4 TRAY 10x6 ARRAY 2,03 DP H44-260465-66C02 Material: ChipSentry (Black Conductive Polycarbonate) Pocket Dimensions: 0.260 x 0.465 x 0.080 Capacity: 10x6 (60) 1 * 200 ST
PRODUKT
Erweitert / Kompakt
4 TRAY 10x6 ARRAY 2,03 DP H44-260465-66C02 Material: ChipSentry® (Black Conductive Polycarbonate) Pocket Dimensions: 0.260´ x 0.465´ x 0.080´ Capacity: 10x6 (60) 1 * 50 ST ENTEH4426046566C02 1 * 50 ST
Hersteller-Artikelnummer:: H4426046566C02
Lokale Artikelnummer:: ENTEH4426046566C02
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder