Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Multiple wafer shipping box for 125 mm (5") wafer handling, Ultrapak® series

Lieferant: ENTEGRIS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

H9125-0028 H9125-0029
ENTEH9125-0028EA 0 EUR InStock
ENTEH9125-0028 ENTEH9125-0029
Multiple wafer shipping box for 125 mm (5") wafer handling, Ultrapak® series
Wafer-Handling-Zubehör Wafer-Versandbehälter
Ultrapak® series multiple wafer shipping boxes are made of Ultrapure polypropylene materials that provide a clean shipping environment and assure low levels of condensable inorganics.

  • Individual horizontal cantilever springs limit wafer rotation to reduce particle contamination and also secure the wafers in the cassette to safeguard against breakage
  • Ultrapure polypropylene materials provide a clean shipping environment and assure low levels of condensable inorganics
  • Horizontal and vertical robotic pickup flanges on the cassette allow convenient handling
  • Center notch track alignment and 'H' bar ensure accurate equipment interoperability both vertically and horizontally
  • Assembly overall size: 161×177×160 mm
  • Cassette overall size: 144×153×146 mm

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge