In-line gauge protector
Lieferant: ENTEGRIS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
IGP-02-060-2-6
ENTEIGP-02-060-2-6EA 0 EUR InStock
ENTEIGP-02-060-2-6
In-line gauge protector
Pumpen Vakuum-Messgeräte
Isolates corrosive chemicals from gauges.
  • All wetted surfaces are of molded PFA material
  • Exterior gauge protector body surfaces are PFA and PVDF material
  • Full flow-through design
  • Gauges consist of a 2 ¹/₂" (63,5 mm) dial, with stainless steel case and stainless steel movement
  • Accepts most ¹/₄" MNPT gauge and transducer designs
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge