Wafer isolation pods, M20X series
Lieferant: ENTEGRIS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M200-5033
ENTEM200-5033EA 0 EUR InStock
ENTEM200-5033
Wafer isolation pods, M20X series
Wafer-Handling-Zubehör Wafer-Carrier
Advanced wafer isolation for 200 mm fabs. Trends in wafer processing technology have mandated advancements in wafer handling technology to support today's advanced semiconductor processing facilities. Advanced wafer isolation pods provide wafer isolation, and coupled with advanced wafer transport carriers, offer performance benefits over traditional wafer carriers and storage boxes. The M20X Series 200 mm Pod is a component of Silicon Delivery™ Systems and Services – Entegris's unique materials integrity management solution to cost effectively manage silicon from production to consumption.
  • Accommodate most 25 and 26 capacity 200 mm wafer carriers
  • Compliant with SEMI® standard E19
  • Side rails for PGV and AGV transport
  • Side handles for manual handling
  • Inert gas purge compatible option
  • Compatible with multiple ID options
  • Cleaned and cleanroom packaged
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge