Complement C3a des Arg (human), ELISA kit
Lieferant: ENZO LIFE SCIENCES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ADI900058
ENZOADI900058EA 776 EUR InStock
ENZOADI900058
Complement C3a des Arg (human), ELISA kit
Tests ELISAs
For the quantitative determination of Complement C3a des arg in human EDTA plasma.
The Complement C3a des Arg (human), EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. This kit comes ready-to-use with sample preparation reagents included. The Complement C3a des Arg (human), EIA kit is specific, offering low cross-reactivity to structurally related molecules, and sensitive with quantification of as little as 120pg/ml Complement C3a des Arg.
Bestellinformation: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer 10 concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Complement reagent A, Complement reagent B.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge