LLE High Recovery Vial Racks and Vials
Lieferant: EPREP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
01-03126
EPRE01-03126EA 55 EUR InStock
EPRE01-03126
LLE High Recovery Vial Racks and Vials
Instrumente zur Probenvorbereitung
ePrep 20 and 50 ml high recovery PP vials are ideal for syringe LLE techniques (Patent Pending) on the ePrep Sample Preparation Workstation.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge