Screw caps
Lieferant: EP SCIENTIFIC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
067-3800WBP 067-4800WBP 067-2410WBP
EPSC067-3800WBPEA 364 EUR InStock
EPSC067-3800WBP EPSC067-4800WBP EPSC067-2410WBP
Screw caps
Verschlüsse Flaschenverschlüsse
LDPE-lined white caps.
  • For general-purpose HDPE bulk separate containers
  • Same high quality caps used on certified HDPE environmental sample containers
Bestellinformation: Bottles sold separately (IR013-series and IR012-series).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge