Jars, clear or amber Type III soda-lime glass, with PTFE-lined white polypropylene closed-top cap, designed for collecting soil, sludge, waste and water samples for a variety of organic and inorganic target analyses.
- Certified to meet EPA performance-based specifications for semi-volatile organics, pesticides, PCBs and metals analyses
- Processed the same way as the certified jars but without certification documentation
- Unprocessed and ready for your own cleaning procedure
- Closures assembled on bottles
- Available as short or tall profile
Lieferumfang: Certified jars supplied with Certificate of Analysis (CofA) in the case and barcoded with lot number for traceability.