Closed caps, PTFE lined
Lieferant: EP SCIENTIFIC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
B267-2414UBB
EPSCB267-2414UBBEA 126 EUR InStock
EPSCB267-2414UBB
Closed caps, PTFE lined
Verschlüsse Flaschenverschlüsse
These PTFE-lined closed caps are replacement caps for Thermo Scientific glass environmental sample bottles, jars, jugs and vials.
The PTFE liner is bonded to the cap so it won't fall out or become separated from the cap.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge