Restriction enzymes, BamHI, HC, Fermentas
Lieferant: FERMENTAS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ER0053
ER0053EA 144 EUR InStock
ER0053
Restriction enzymes, BamHI, HC, Fermentas
Nukleinsäure-Reagenzien Restriktionsenzym
BamHI restriction enzyme recognises G^GATCC sites and cuts best at 37 °C in its own unique buffer.
  • Superior quality, stringent quality control and industry leading manufacturing process
  • Convenient colour-coded five buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge