Restriction enzymes, EcoRI, Fermentas
Lieferant: FERMENTAS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ER0271 ER0272 ER0275
ER0271EA 49.4 EUR InStock
ER0271 ER0272 ER0275
Restriction enzymes, EcoRI, Fermentas
Nukleinsäure-Reagenzien Restriktionsenzym
These EcoRI restriction enzyme recognizes G^AATTC sites and cuts best at 37 °C in its own unique buffer. Exhibit 100% activity in the recommended buffer and reaction conditions.
  • Superior quality, stringent quality control and industry leading manufacturing process
  • Convenient color-coded five buffer system and BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that sent in DNA preparations. Includes universal Tango buffer for double-digestions.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge