Ergonomic mat, Classic with yellow bevels
Lieferant: ERGOMAT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ST-RUNNER
ERGOST-RUNNEREA 670 EUR InStock
ERGOST-RUNNER
Ergonomic mat, Classic with yellow bevels
Fußmatten Anti-Ermüdungsmatten
The Classic is the original ergonomic bubble profile mat. Customers still have many of these original mats in service for 20 years or longer.
  • Ergonomic
  • Made of foamed polyurethane
  • Beveled edges
  • Anti-static and silicon free
  • Any size, any shape
  • True warranty
Designed to stimulate circulation in feet, legs and lower back. Circulation is stimulated by the unique, patented surface profile of Ergomat mats, which forces muscular response. Molded with bevels. Yellow bevels can be added. Bevels will never curl up, eliminating trip hazards.
Ideal for environments requiring static control. Can be custom made in any size and shape, creating one continuous surface to eliminate seams and trip hazards. All Ergomat mats are silicon free.
Zertifizierungen: Classic mats are ISO cleanroom Class 5 (Class 100) rated.
Bestellinformation: Eight-year warranty. Ergomat stands 100% behind its products with full replacement coverage if the product fails to perform as promoted within that period. At Ergomat this is called a True Warranty.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge