High Speed Camera, CMEX-18 Pro
Lieferant: EUROMEX
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
DC.18000-PRO
EUMXDC.18000-PROEA 1210 EUR InStock
EUMXDC.18000-PRO
High Speed Camera, CMEX-18 Pro
Kameras
High speed camera with perfect colour-reproduction capability.
  • 18,0 Mpix CMOS colour USB 3,0 camera
  • 1/2,3" sensor, 4912×3684 pixels
  • Low signal/noise ratio
  • USB 3.0 interface (USB 2.0 compatible)
Housing: Integrated CNC aluminum alloy
Power supply: 5 V over PC USB port
Lieferumfang: Supplied with 0,5× objective with C-mount, USB 3 cable, 30 and 30,5 mm adapters for stereomicroscopes, 76×24 mm calibration slide (1 mm/100), carton box.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge