Conductivity/ATC electrodes, 2-cell, Eutech
Lieferant: EUTECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
CONSEN91W CONSEN91B CONSEN9201D
EUTECONSEN91WEA 242 EUR InStock
EUTECONSEN91W EUTECONSEN91B EUTECONSEN9201D
Conductivity/ATC electrodes, 2-cell, Eutech
Elektroden Leitfähigkeitselektroden
Laboratory electrodes for measuring conductivity and temperature.
  • Stainless steel rings, PP and ultem-body
  • 6-pin, 8 pin or BNC connector
  • For use with different Eutech meters
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge