Conductivity meter, bench top, Eutech CON 2700
Lieferant: EUTECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ECCON270043S
EUTEECCON270043SEA 1170 EUR InStock
EUTEECCON270043S
Conductivity meter, bench top, Eutech CON 2700
Elektrochemische Messgeräte Leitfähigkeitsmessgeräte
The Eutech CON 2700 accepts 2-cell and 4-cell electrodes, allowing a broad range of measurement at up to ±1 % full scale accuracy. It can measure conductivity, TDS, salinity, resistivity and the temperature.
  • Multi data screen shows conductivity readings, temperature, electrode status, calibration points, date and time
  • Auto-calibration with preset values for quick easy calibration, or manual multi-point calibration for greater accuracy
  • Quick, easy electrode diagnosis with the effective cell constants display
  • Non-volatile memory holds up to 500 data points – time and date-stamped for GLP compliance
  • Bi-directional RS232 for easy data transfer to computer
  • Cal-due alarm – no more out-dated calibrations
  • Limit alarm alerts when reading falls out of range
  • Password protection for setup and calibration
Typical applications include: Environmental studies, chemical laboratories, quality assurance testing, food science, ecological studies, education.
Lieferumfang: Supplied with 4-cell conductivity/ATC probe (CONSEN9201D), RS232 cable, 100/240 VAC power adapter and an integral electrode holder.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge