Ion selective meter, bench top, Eutech ION 2700
Lieferant: EUTECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ECION270040S ECION270042GS
EUTEECION270040SEA 1210 EUR InStock
EUTEECION270040S EUTEECION270042GS
Ion selective meter, bench top, Eutech ION 2700
Elektrochemische Messgeräte Ionen-Messgeräte
This bench meter comes with a large, one-glance-sees-all screen. Users can view pH, ION or Redox readings together with temperature, electrode status, calibration points, date and time all at once.
  • Up to 6-point push button calibration with auto-buffer recognition
  • Quick, easy electrode diagnosis with pH slope and offset display
  • Non volatile memory holds up to 500 data points – time and date-stamped for GLP compliance
  • Bi-directional RS232 for easy data transfer to computer
  • Cal-due alarm – no more out-dated calibrations
  • Auto-logging function for convenient continuous monitoring
  • Splashproof housing and keypad protects against incidental spills
Lieferumfang: Supplied with RS232 cable, 100/240 VAC power adapter and an integral electrode holder.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge