Electrode stand with swivel arm, Eutech
Lieferant: EUTECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ECPHELSTDC
EUTEECPHELSTDCEA 123 EUR InStock
EUTEECPHELSTDC
Electrode stand with swivel arm, Eutech
Elektrodenhalter
This electrode stand with swivel arm allows convenient hands-free operation. Holds up to four standard size electrodes and one ATC probe.
  • Weighted 6-inch diameter base allows users to pivot holder 360° for more flexibility
  • Arm allows users to move electrode side to side or up and down while keeping electrode at a constant vertical angle
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge