pH/mV/°C meters, handheld, Eutech CyberScan pH 600 / 610 / 620
Lieferant: EUTECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ECPHWP61042K ECPHWP60001 ECPHWP60042K ECPHWP61001 ECPHWP62042K
EUTEECPHWP61042KEA 1100 EUR InStock
EUTEECPHWP61042K EUTEECPHWP60001 EUTEECPHWP60042K EUTEECPHWP61001 EUTEECPHWP62042K
pH/mV/°C meters, handheld, Eutech CyberScan pH 600 / 610 / 620
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
The CyberScan pH 600 series enables a wireless and cable-free data transfer with infrared IrDA technology. Data can be simply sent from the meter to a PC with the press of a button.
  • Large screen with backlight displays electrode status, calibration information, temperature and pH or ion measurements
  • Memory stores/ recalls up to 500 data sets with temperature readings, with time and date-stamping formatted to meet GLP standards
  • 6 calibrations points and 20 auto/manual buffer options sets for quick, convenient calibration
  • Alerts when probe requires maintenance with pH probe quality indicator average slope
  • Rugged and waterproof to IP67 standard with protective rubber boot and handy velcro strap for better grip
  • Self-diagnostic with message codes for easy troubleshooting
  • Ion-measurement capabilities with up to 8 Ion calibration points (pH 620 only)
pH 600 and pH 610 measure pH, mV and temperature while the pH 620 additionally measures ion concentrations.
Ideal for testing the water quality, monitoring the health of aquatic ecosystems, and for surveying surface and ground water drinking supplies. Also suitable for checking the quality of plant water intake and discharge, wastewater and water treatment, recirculating systems, industrial process systems, fish farming, water conditioning plants and chemical process verification.
Lieferumfang: Supplied with PC-based complimentary Data Acquisition Software for hassle-free data import and report-generation into any popular spreadsheet program.
Some kits are supplied with the pH carrying kit set incl. a plastic carrying case, 1× pH 4,01, pH 7,00 buffer solutions, storage solution and 1× deionised (rinse) water (60 ml each).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge