pH tester, Eutech pHTestr 10 BNC
Lieferant: EUTECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PHTEST10BNC
EUTEPHTEST10BNCEA 195 EUR InStock
EUTEPHTEST10BNC
pH tester, Eutech pHTestr 10 BNC
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
This water- and dust-proof tester with BNC connection is ideal for quick and accurate pH checks in pools and spas, aquariums and hydroponics operations, cooling towers, food processing and water quality testing.
  • Large dual display for enhanced readability, provides comprehensive meter information
  • Water and dust-proof housing (IP67 rated): Meter can float
  • User-selectable electrodes with BNC connectors - for greater flexibility with choice of electrodes for specific applications
  • Push-button calibration with auto-buffer recognition - quick, easy calibration with no calibration mistakes
  • Auto power-off saves battery power after non use
  • "Hold" function freezes reading momentarily for easy viewing
  • Manual Temperature Compensation (MTC)
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge