Anti-PLA2G4C Goat Polyclonal Antibody
Lieferant: Everest Biotech
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
EB10114
EVBIEB10114EA 339 EUR InStock
EVBIEB10114
Anti-PLA2G4C Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PLA2G4C (phospholipase A2, group IVC (cytosolic, calcium-independent))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge