Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SCARB1 Goat Polyclonal Antibody

Lieferant: Everest Biotech

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

EB12300
EVBIEB12300EA 339 EUR InStock
EVBIEB12300
Anti-SCARB1 Goat Polyclonal Antibody
Antikörper
Western Blot: Approx 85kDa band observed in Human Adrenal gland lysates (calculated MW of 57.0kDa according to NP_005496.4). We do note that SCARB1 is a glycoprotein which makes a higher size band to be expected, and this is consistent with observations

Type: Primary
Antigen: SCARB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge