Avantor® ACE® Excel® Oligo, HPLC/UHPLC Columns, Analytical
Lieferant: Advanced Chromatography Technologies
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
EXL-1715-0502EA 937 EUR InStock
EXL-1715-0502 EXL-1715-1002 EXL-1115-0502 EXL-1115-1002 EXL-1115-1502 EXL-1115-1046 EXL-1115-1546 EXL-1P15-GD2U EXL-1P15-GD4U
Avantor® ACE® Excel® Oligo, HPLC/UHPLC Columns, Analytical
Chromatographiesäulen
Avantor® ACE® Oligo is a unique encapsulated stationary phase for high resolution ion-pair reversed-phase analysis of oligonucleotides. These high pH stable stainless steel columns are compatible with all buffers and mobile phases typically used for oligonucletide analysis by LC-UV and LC-MS, including TEAA and HFIP.
  • High efficiency 1,7 and 3,0 µm particle sizes for high resolution oligonucleotide separations
  • Ultra-inert, base deactivated silica for exceptional reproducibility and peak shape
  • Excellent column lifetime under oligonucleotide testing conditions
  • Ultra-low bleed for excellent LC/MS compatibility
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge