Aqueous mounting medium, Fluoroshield™ with PI, Sigma-Aldrich®
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F5932-20ML
F5932-20MLEA 102 EUR InStock
F5932-20ML
Aqueous mounting medium, Fluoroshield™ with PI, Sigma-Aldrich®
Färbemittel Mikroskopie- und Färbereagenzien
Fluoroshield™ with PI is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.
  • Storage temperature: 2 - 8 °C
This mounting medium is fortified with propidium iodide (PI) which is a counterstain for DNA. For use in in situ hybridization techniques or other methods where fluorescence of DNA staining is required. PI excites at 535 nm and emits at 615 nm, producing a red fluorescence. RNA is also stained with PI.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge