Aqueous mounting medium, Fluoroshield™ with PPD, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F6812-20ML
F6812-20MLEA 97.5 EUR InStock
F6812-20ML
Aqueous mounting medium, Fluoroshield™ with PPD, Sigma-Aldrich®
Färbemittel Mikroskopie- und Färbereagenzien
Fluoroshield™ with PPD (p-phenylenediamine; 1,4-phenylenediamine) is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.
  • Storage temperature: 2 - 8 °C
Fluoroshield prevents rapid photobleaching of FITC, Texas Red®, AMCA, Alexa Fluor® 488, Alexa Fluor 594, and tetramethylrhodamine. This mounting medium is fortified with PPD, an anti-fade reagent used to retard photobleaching of fluorescein and other fluorescent dyes. NOTE: Fluoroshield with PPD is NOT SUITABLE for immunofluorescence techniques involving Cy® dyes, R-phycoerythrin (R-PE), phycocyanin (PC),or allophycocyanin (APC).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge