Restriction enzymes, XhoI, FastDigest™, Fermentas
Lieferant: FERMENTAS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
FD0694 FD0695
FD0694EA 63.25 EUR InStock
FD0694 FD0695
Restriction enzymes, XhoI, FastDigest™, Fermentas
Nukleinsäure-Reagenzien Restriktionsenzym
FastDigest™ XhoI is one of an advanced line of fast restriction enzymes that are all 100% active in the universal FastDigest™ and FastDigest™ Green reaction buffers.
  • 100% buffer compatibility with downstream applications
  • Complete digestion in 5 to 15 minutes
  • Direct loading on gels
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge