Scalpels, disposable, Feather®
Lieferant: FEATHER
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
200130010 200130023 200130024 200130022 200130021 200130020 200130015 200130011 200130025 02.00210015
FEAT200130010EA 23 EUR InStock
FEAT200130010 FEAT200130023 FEAT200130024 FEAT200130022 FEAT200130021 FEAT200130020 FEAT200130015 FEAT200130011 FEAT200130025 FEAT02.00210015
Scalpels, disposable, Feather®
Skalpelle Skalpelle, komplett
These disposable standard scalpels with high-quality stainless steel blade are suitable for all surgical applications.
  • Outstanding quality and performance thanks to optimised micrometer-accurate precision grinding
  • Double-honing process ensures ultra-sharp cutting edges of great durability
Verpackung: 20 pieces per pack.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge