Autopsy blade handles and blades, FEATHER®
Lieferant: FEATHER
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
205000170 205000100 205000000 205000325
FEAT205000170EA 210 EUR InStock
FEAT205000170 FEAT205000100 FEAT205000000 FEAT205000325
Autopsy blade handles and blades, FEATHER®
Mikrotome und Mikrotomanordnungen Mikrotomklingen und -griffe
The plastic handle is autoclavable and features excellent chemical resistance. It can be easily disassembled for cleaning. Superior sharpness and cutting performance compared to conventional knives. The same handle can be used for brain dissection and other purposes simply by exchanging the blade.
  • Fast and easy blade change
  • Handle is easy to clean and autoclavable
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge