Vacuum wand, C-series
Lieferant: WINDRUSH TECHNOLOGY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C003-Y-98-CP 106518 90-CP C002-Y-92-CP C002-Y-90-CP C003-X-95-CP 98-CP C003-Y-99-CP C003-Y-91-CP C002-X-95-CP C003-Y-92-CP C002-Y-98-CP C003-X-96-CP C001-Y-91-CP C002-X-96-CP 99-CP 91-CP C002-Y C003-Y-90-CP
FLUMC003-Y-98-CPEA 0 EUR InStock
FLUMC003-Y-98-CP FLUM106518 FLUM90-CP FLUMC002-Y-92-CP FLUMC002-Y-90-CP FLUMC003-X-95-CP FLUM98-CP FLUMC003-Y-99-CP FLUMC003-Y-91-CP FLUMC002-X-95-CP FLUMC003-Y-92-CP FLUMC002-Y-98-CP FLUMC003-X-96-CP FLUMC001-Y-91-CP FLUMC002-X-96-CP FLUM99-CP FLUM91-CP FLUMC002-Y FLUMC003-Y-90-CP
Vacuum wand, C-series
Vakuum-Systeme
This C series wand bodies are manufactured in ESD safe materials including PEEK tips are the safest means of wafer handling. The tips are optimally sized for each wafer diameter, ensuring that contact is both minimal and secure.
  • Polished contact surface provides excellent adhesion to water
  • Fixed tip and swivel joint body styles are available
  • Unique control valve ensures reliable suction and instantaneous release without particle generation
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge