Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Ball mills, mini-mill PULVERISETTE 23

Lieferant: FRITSCH

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

23.1000.00
FRIT23.1000.00EA 4020 EUR InStock
FRIT23.1000.00
Ball mills, mini-mill PULVERISETTE 23
Mühlen
Ball mill for fine comminution of minimal quantities of dry laboratory samples or solids in suspensions. Can also be used for mixing and homogenisation of emulsions.

  • Small grinding bowl volume for minimal sample quantities <5 ml
  • Small contact surface with grinding tools
  • Maintenance-friendly with quick grinding bowl fastening with tongue and groove clamping system
  • Oscillation amplitude 9 mm
  • Adjustable, reproducible grinding duration and adjustable loading frequency

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge