Silver neuro clips
Lieferant: FINE SCIENCE TOOLS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
18040-00
FSCI18040-00EA 137 EUR InStock
FSCI18040-00
Silver neuro clips
Erste-Hilfe-Ausstattung Wundklammern
These single-use silver neuro clips are meant for permanent vessel occlusion.
  • Clamp Press: Variable
  • Alloy / Material: Silver
  • Length: 5 mm
  • Not serrated
Achtung! Bitte beachten: All surgical instruments and devices from the Fine Science Tools product range are intended exclusively for use in experimental research laboratories and facilities or in veterinary medicine.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge