Organic reference standard, beta-D-Glucan from barley, for use with Total Dietary Fiber Control Kit, TDF-C10, Supelco®
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
G7391-1G
G7391-1GEA 482 EUR InStock
G7391-1G
Organic reference standard, beta-D-Glucan from barley, for use with Total Dietary Fiber Control Kit, TDF-C10, Supelco®
Standards Organische Standards
β-D-Glucan is a β-D-glucose polysaccharide, naturally found in the cell wall of fungi, bacteria and plants.
Internal control for use with the Total Dietary Fiber Assay Kit.
Verpackung: 1 g in poly bottle
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge