Glucose (HK) assay kit
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
GAHK20-1KT
GAHK20-1KTEA 107 EUR InStock
GAHK20-1KT
Glucose (HK) assay kit
Tests Zelltests
The Glucose (HK) Assay Kit is for the quantitative, enzymatic determination of glucose in food and other material.
Glucose is phosphorylated by adenosine triphosphate (ATP) in the reaction catalysed by hexokinase. Glucose-6-phosphate (G6P) is then oxidised to 6-phospho-gluconate in the presence of oxidised nicotinamide adenine dinucleotide (NAD) in a reaction catalysed by glucose-6-phosphate dehydrogenase (G6PDH). During this oxidation, an equimolar amount of NAD is reduced to NADH. The consequent increase in absorbance at 340 nm is directly proportional to glucose concentration.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge