Autoclave-Resistant Labels for Laser Printers (A4 size)
Lieferant: GA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
A4AKA-23WH A4AKA-3WH A4AKA-8WH
GAINA4AKA-23WHEA 86.4 EUR InStock
GAINA4AKA-23WH GAINA4AKA-3WH GAINA4AKA-8WH
Autoclave-Resistant Labels for Laser Printers (A4 size)
Etiketten
These laser labels have a permanent adhesive, perfect for identifying disposable sterilisation pouches and bags. Provided in A4 sheet format for printing with most common desktop laser printers, they can be printed with human-readable text, serialised numbering, as well as 1D/2D barcodes.
  • Temperature range: −40 to 150 °C (−40 to 302 °F)
  • Strong permanent adhesive ensures labels adhere to glass, metal, and plastic surfaces
  • Designed for use in steam autoclaves, dry heat ovens, and with gamma irradiators (up to 50 KGy) and ethylene oxide sterilisers
  • Compatible with all major brands of desktop laser printers
  • Water-proof and alcohol-resistant
Permanent laser printable labels, ideal for glassware, surgical instruments, metal and plastic surfaces, trays, blister packs, and other items intended for use in steam autoclaves, ovens, and gamma irradiators. These autoclave laser labels are durable, inert, weather- and water-proof, alcohol-resistant, and have been tested to endure up to 20 autoclave cycles (at 121 °C/250 °F, 16,5 psi, for 20 minutes). Compatible with all major brands of desktop laser printers.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge