Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

DYMO-compatible cryogenic labels

Lieferant: GA

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

EG1F-072BE EG1F-072BL EG1F-072BR EG1F-072GA EG1F-072GR EG1F-072LE EG1F-072OR EG1F-072RE EG1F-072VI EG1F-072YE EG1F-081BE EG1F-081BL EG1F-081BR EG1F-081GA EG1F-081GR EG1F-081LE EG1F-081OR EG1F-081RE EG1F-081VI EG1F-081YE
GAINEG1F-072BEEA 90.6 EUR InStock
GAINEG1F-072BE GAINEG1F-072BL GAINEG1F-072BR GAINEG1F-072GA GAINEG1F-072GR GAINEG1F-072LE GAINEG1F-072OR GAINEG1F-072RE GAINEG1F-072VI GAINEG1F-072YE GAINEG1F-081BE GAINEG1F-081BL GAINEG1F-081BR GAINEG1F-081GA GAINEG1F-081GR GAINEG1F-081LE GAINEG1F-081OR GAINEG1F-081RE GAINEG1F-081VI GAINEG1F-081YE
DYMO-compatible cryogenic labels
Etiketten
DYMO compatible, direct thermal cryo labels. For ultra-low and cryogenic temperature applications as low as –196 °C.

  • EG1F-081 series labels are resistant to gamma irradiation (tested to up to 50 kGy)
  • Ideal for the identification of 0,5 to 5 ml vials and tubes
  • Compatible with all DYMO label writers
  • Temperature range: –196 to 70 °C
  • Permanent adhesive

Ideal for identification and permanent storage of vials and containers in liquid nitrogen and laboratory freezers (–80, –50, –20 °C). Labels will also resist extreme cryogenic temperatures (–196 °C in a liquid phase and vapor phase liquid nitrogen).

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge