Deep-Freeze Removable Tapes
Lieferant: GA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
TRM-13C1-50WH TRM-13C1-50BL TRM-13C1-50GA TRM-13C1-50OR TRM-13C1-50RE TRM-13C1-50YE TRM-13C3-100WH TRM-13C3-100BL TRM-13C3-100GA TRM-13C3-100OR TRM-13C3-100RE TRM-13C3-100YE TRM19SBC1-50WH TRM19SBC1-50BL TRM19SBC1-50GA TRM19SBC1-50OR TRM19SBC1-50RE TRM19SBC1-50YE TRM19SBC3100WH TRM19SBC3100BL TRM19SBC3100GA TRM19SBC3100OR TRM19SBC3100RE TRM19SBC3100YE TRM25SBC1-50WH TRM25SBC1-50BL TRM25SBC1-50GA TRM25SBC1-50OR TRM25SBC1-50RE TRM25SBC1-50YE TRM25SBC3100WH TRM25SBC3100BL TRM25SBC3100GA TRM25SBC3100OR TRM25SBC3100RE TRM25SBC3100YE
GAINTRM-13C1-50WHEA 27.7 EUR InStock
GAINTRM-13C1-50WH GAINTRM-13C1-50BL GAINTRM-13C1-50GA GAINTRM-13C1-50OR GAINTRM-13C1-50RE GAINTRM-13C1-50YE GAINTRM-13C3-100WH GAINTRM-13C3-100BL GAINTRM-13C3-100GA GAINTRM-13C3-100OR GAINTRM-13C3-100RE GAINTRM-13C3-100YE GAINTRM19SBC1-50WH GAINTRM19SBC1-50BL GAINTRM19SBC1-50GA GAINTRM19SBC1-50OR GAINTRM19SBC1-50RE GAINTRM19SBC1-50YE GAINTRM19SBC3100WH GAINTRM19SBC3100BL GAINTRM19SBC3100GA GAINTRM19SBC3100OR GAINTRM19SBC3100RE GAINTRM19SBC3100YE GAINTRM25SBC1-50WH GAINTRM25SBC1-50BL GAINTRM25SBC1-50GA GAINTRM25SBC1-50OR GAINTRM25SBC1-50RE GAINTRM25SBC1-50YE GAINTRM25SBC3100WH GAINTRM25SBC3100BL GAINTRM25SBC3100GA GAINTRM25SBC3100OR GAINTRM25SBC3100RE GAINTRM25SBC3100YE
Deep-Freeze Removable Tapes
Etiketten
Removable lab tape for deep-freeze storage of cryo containers, including freezer boxes, tubes, vials, plastic bags, and other items intended for long-term storage in laboratory freezers and dry ice transportation. Writable using permanent cryo markers.
  • Temperature range: −80 to 100 °C (−112 to 212 °F)
  • Resists long-term storage in deep-freeze conditions, including multiple freeze-thaw cycles
  • Glove-friendly and removable; won't leave behind any adhesive residue
  • Available in a range of colors for easy color coding
Deep-freeze removable tape for sealing or identifying cryo and freezer boxes, tubes, vials, plastic bags, and other items intended for long-term storage in laboratory freezers (−20 °C, −40 °C, −80 °C) and for dry ice transportation. Writable with permanent cryo markers, glove-friendly, withstands dry ice, multiple freeze-thaw cycles, gamma irradiation, and other harsh laboratory environments. The tape should be applied at room temperature before storage in freezers. Not recommended for use in liquid nitrogen.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge