Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Ampicillin, HP Ampillicin™

Lieferant: G Bioscienes

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


GD-6060
GD-6060EA 418 EUR InStock
GD-6060
Ampicillin, HP Ampillicin™
Ampicillin
HP Ampicillin™ is a stronger and more stable inhibitor of ß-lactamase than regular ampicillin and therefore supports only the growth of ampicillin resistant colonies even after prolonged incubation. Culture plates can be incubated for a longer period at 37 °C, eliminating the time constraint associated with normal ampicillin.

  • Reduces satellite colonies
  • Lyophilised format for improved stability
  • Reconstituted 1000X concentration
  • Suitable for 5 L media or agar

Lieferumfang: HP Ampicillin™ is supplied lyophilised. When reconstituted in 500 µl deionised water it produces a 1000X solution. Each kit contains 10 vials, suitable for a total volume of 5 L culture media or agar.


Formel: C₁₆H₁₉N₃O₄S
MW: 349.4 g/mol
MDL Number: MFCD00005175
CAS-Nummer: 69-53-4
Order Now

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge