Affinity chromatography columns, HiScreen™ IXSelect
Lieferant: Cytiva
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
17-3714-10
GEHE17-3714-10EA 1356 EUR InStock
GEHE17-3714-10
Affinity chromatography columns, HiScreen™ IXSelect
Chromatographiesäulen
HiScreen™ IXSelect columns are prepacked with IXSelect affinity chromatography medium. These columns are an excellent choice for method optimisation and parameter screening.
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography resin using the same linear fluid velocity
IXSelect is a high-throughput resin specifically designed for the purification of factor IX targets from recombinant feed streams.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge