Human Serum AB (Off The Clot), Gamma Irradiated
Lieferant: GEMINI BIO-PRODUCTS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
100-318G-100
GEMB100-318G-100EA 923 EUR InStock
GEMB100-318G-100
Human Serum AB (Off The Clot), Gamma Irradiated
Seren
Off the clot human serum AB is collected from normal healthy male donors at FDA-licensed facilities in the United States. As a natural off the clot product, this material contains no anticoagulants/preservatives. All donor units are tested for viral markers and found to be non-reactive.
  • Supports clinical and commercial applications
  • Aseptic assurance system
  • Gamma irradiated
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge