T4 UvsX DNA Recombinase
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-1989 786-1990
GENO786-1989EA 498 EUR InStock
GENO786-1989 GENO786-1990
T4 UvsX DNA Recombinase
Nukleinsäure-Reagenzien Modifikation und Markierung von Nukleinsäure
T4 UvsX DNA recombinase mediates DNA strand exchange between homologous chromosomes. The protein forms a right-handed nucleoprotein complex on single ssDNA called the presynaptic filament that can search for homology in duplex DNA and pair the recombining DNA molecules to form a DNA joint molecule. This process is aided by ssDNA binding proteins and recombination mediators. The filament is then elongated by DNA polymerase with the newly synthesised strand displacing the old strand. The newly synthesised duplex DNA can then act as a template for the next cycle to achieve exponential amplification of dsDNA. E.coli cells expressing recombinant T4 UvsX gene.
  • UvsX DNA recombinase is free from detectable nuclease contamination
Verpackung: Product includes T4 UvsX DNA recombinase 10X UvsX DNA recombinase reaction buffer.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge