picoLUCENT™ PLUS-HRP Kit
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-09 786-002 786-09T 786-424 786-264 786-165
786-09EA 256 EUR InStock
786-09 786-002 786-09T GENO786-424 GENO786-264 GENO786-165
picoLUCENT™ PLUS-HRP Kit
Elektrophorese-Reagenzien Western Blotting Reagents
The picoLUCENT™ PLUS-HRP kit is based on an ultra sensitive luminol substrate that produces chemiluminescence upon reaction with horseradish peroxidase. The chemiluminescence light emission can be recorded by a short exposure to autoradiography films.
  • Low picogram detection (10⁻¹²) of antigens
  • Low noise (signal/background ratio)
Lieferumfang: Complete kit includes a non animal protein blocking agent (NAP-BLOCKER™) and wash buffer (femto-TBST™) and contains sufficient reagents for 25 mini blots (8×7,5 cm) or 1,500 cm² of PVDF or nitrocellulose membrane.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge