ELISA chemiluminescent detection system, OptiBlaze™
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-302 786-539
786-302EA 909 EUR InStock
786-302 GENO786-539
ELISA chemiluminescent detection system, OptiBlaze™
Tests ELISAs
OptiBlaze™ ELISA femto-HRP and femto-AP are stabilised ultra sensitive luminol and 1,2 dioxetane based horseradish peroxidase or alkaline phosphatase substrates for the detection of HRP or AP-conjugated antibodies during ELISA.
OptiBlaze™ kits are chemiluminescent detection systems for Enzyme-Linked Immunosorbent Assay (ELISA). The chemiluminescent substrates provided in the kit are ultra sensitive substrates developed for luminometer-based applications, specific for horseradish peroxidase or alkaline phosphatase labelled antibodies.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge