DNA ladder, 100 bp, DNAmark™
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-855
GENO786-855EA 163 EUR InStock
GENO786-855
DNA ladder, 100 bp, DNAmark™
Elektrophorese-Reagenzien Molekulargewichtsmarker
DNAmark™ 100 bp ladder consists of 11 fragments ranging from 100 to 1500 base pairs (bp). The 500 bp fragment is present at increased intensity to allow easy identification. All fragments are precisely quantified and mixed during the production. For 5 µl loading, all fragments except 500 bp are 40 ng. The 500 bp fragment is 100 ng.
  • Pre-mixed with loading dye and ready to use
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge