GENTAMICIN USP GRADE 10G 1 * 10 g
Lieferant: G Bioscienes
Gentamicin efficiently inhibits growth of many Gram-positive and Gram-negative microorganisms. It does so by inhibiting protein synt hesis by binding to the L6 protein of the 30S ribosomal subunit. It is also effective against mycoplasma. Recommended 1 * 10 g
PRODUKT
Erweitert / Kompakt
GENTAMICIN USP GRADE 10G 1 * 10 g GENORC-173 1 * 10 g
Hersteller-Artikelnummer:: RC-173
Lokale Artikelnummer:: GENORC-173
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder