Hydrometer for yogurt and chocolate milk
Lieferant: GERBER FUNKE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
6670
GERB6670EA 21.2 EUR InStock
GERB6670
Hydrometer for yogurt and chocolate milk
Aräometer
Hydrometer for yoghurt and chocolate milk, in the body reading at the top, with thermometer.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge