Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Nitrogen/protein analysers, Dumatherm®

Lieferant: GERHARDT

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

14-0500 14-0400
GERH14-0500EA 0 EUR InStock
GERH14-0500 701-0261
Nitrogen/protein analysers, Dumatherm®
Analysatoren Protein-Analysatoren
For the analysis of solid and liquid samples in Dumatherm®, the sample is burned at high temperatures, followed by an analysis of the obtained gases.

  • Detection limit: 0,01 mg N absolute, absolute C 0,02 mg
  • Standard deviation up to 0,3% absolute with 150 mg EDTA
  • Automatic sample processing
  • Safe analysis and increased reproducibility productivity
  • Convenient control software

Determination of proteins in milk and dairy products, cereals, eggs and their products, meat and derivatives, malt and brewery raw materials, animal feed, beer, wine, brewery grain, grain products, oilseeds, starch, and many other food ingredients.

Determination of nitrogen in the soil, water, soil additives and growing media, fertilisers, urine, pulp, paper, petroleum, plastics, explosives, coffee, tobacco.

For further discussion of the carbon level, such as soil samples or mineral oils, we recommend the version Dumatherm® CN.

For sample weight of 0,5 mg to 1 g, the scan takes 2 to 4 minutes for nitrogen and 9 to 12 minutes for the parameters Carbon and Nitrogen.

Zertifizierungen: According to national and international standards and analytical procedures such as § 64 LFGB, DIN, AOAC, DIN EN ISO and Gafta.

Bestellinformation: Oxygen, helium and compressed air or nitrogen gas supplies are required - A control PC is also required. Please contact your Avantor local agency for specifications of gases and requirements of the PC. Comes complete including all consumables and accessories for about 1000 samples.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge