Sani-Tech® ultra-pure platinum-cured silicone tubing meets the stringent requirements of the biopharmaceutical market.
- Maximum temperature range: –62 to +260 °C (–80 to +500 °F)
- Imparts no taste or odour to critical stream
- Available in 50 and 65 durometers
- Laser etched for lot traceability
It features ultra low extractables, excellent flexibility, longer pump life and non-yellowing formulation.
Applications include sterile fill and transfers, drug delivery, cell harvest, media processing, sanitary peristaltic pump tubing, production filtration and fermentation, bioreactor process lines.
Zertifizierungen: Meets USP Class VI and EP 3.1.9 silicone elastomer. Manufactured in an FDA registered and ISO criteria.
Bestellinformation: Available in three different lengths.