Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Tubing, silicone, Sani-Tech®, ULTRA-C Series

Lieferant: SAINT GOBAIN

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

ULTRA-C-012-0 ULTRA-C-020-0 ULTRA-C-025-0 ULTRA-C-030-0 ULTRA-C-030-2 ULTRA-C-040-0 ULTRA-C-058-0 ULTRA-C-062-1 ULTRA-C-062-2 ULTRA-C-062-3 ULTRA-C-062-4 ULTRA-C-062-5 ULTRA-C-078-1 ULTRA-C-078-2 ULTRA-C-078-3 ULTRA-C-078-5 ULTRA-C-093-1 ULTRA-C-093-2 ULTRA-C-093-3 ULTRA-C-093-4 ULTRA-C-1000-4 ULTRA-C-125-1 ULTRA-C-125-2 ULTRA-C-125-3 ULTRA-C-125-4 ULTRA-C-125-5 ULTRA-C-156-4 ULTRA-C-187-1 ULTRA-C-187-2 ULTRA-C-187-3 ULTRA-C-187-4 ULTRA-C-187-5 ULTRA-C-250-1 ULTRA-C-250-2 ULTRA-C-250-3 ULTRA-C-250-4 ULTRA-C-312-1 ULTRA-C-312-2 ULTRA-C-312-3 ULTRA-C-312-4 ULTRA-C-312-5 ULTRA-C-375-2 ULTRA-C-375-3 ULTRA-C-375-4 ULTRA-C-375-5 ULTRA-C-500-2 ULTRA-C-500-3 ULTRA-C-500-4 ULTRA-C-500-5 ULTRA-C-625-2 ULTRA-C-625-4 ULTRA-C-625-5 ULTRA-C-750-4 ULTRA-C-750-5 ULTRA-C-750-6 ULTRA-C-875-2 ULTRA-C-875-4 ULTRA-C-875-5 ULTRA-C-875-6
GESSULTRA-C-012-0EA 46.1 EUR InStock
GESSULTRA-C-012-0 GESSULTRA-C-020-0 GESSULTRA-C-025-0 GESSULTRA-C-030-0 GESSULTRA-C-030-2 GESSULTRA-C-040-0 GESSULTRA-C-058-0 GESSULTRA-C-062-1 GESSULTRA-C-062-2 GESSULTRA-C-062-3 GESSULTRA-C-062-4 GESSULTRA-C-062-5 GESSULTRA-C-078-1 GESSULTRA-C-078-2 GESSULTRA-C-078-3 GESSULTRA-C-078-5 GESSULTRA-C-093-1 GESSULTRA-C-093-2 GESSULTRA-C-093-3 GESSULTRA-C-093-4 GESSULTRA-C-1000-4 GESSULTRA-C-125-1 GESSULTRA-C-125-2 GESSULTRA-C-125-3 GESSULTRA-C-125-4 GESSULTRA-C-125-5 GESSULTRA-C-156-4 GESSULTRA-C-187-1 GESSULTRA-C-187-2 GESSULTRA-C-187-3 GESSULTRA-C-187-4 GESSULTRA-C-187-5 GESSULTRA-C-250-1 GESSULTRA-C-250-2 GESSULTRA-C-250-3 GESSULTRA-C-250-4 GESSULTRA-C-312-1 GESSULTRA-C-312-2 GESSULTRA-C-312-3 GESSULTRA-C-312-4 GESSULTRA-C-312-5 GESSULTRA-C-375-2 GESSULTRA-C-375-3 GESSULTRA-C-375-4 GESSULTRA-C-375-5 GESSULTRA-C-500-2 GESSULTRA-C-500-3 GESSULTRA-C-500-4 GESSULTRA-C-500-5 GESSULTRA-C-625-2 GESSULTRA-C-625-4 GESSULTRA-C-625-5 GESSULTRA-C-750-4 GESSULTRA-C-750-5 GESSULTRA-C-750-6 GESSULTRA-C-875-2 GESSULTRA-C-875-4 GESSULTRA-C-875-5 GESSULTRA-C-875-6
Tubing, silicone, Sani-Tech®, ULTRA-C Series
Schläuche Transferschlauch
Sani-Tech® ultra-pure platinum-cured silicone tubing meets the stringent requirements of the biopharmaceutical market.

  • Maximum temperature range: –62 to 260 °C (–80 to +500 °F)
  • Imparts no taste or odour to critical stream
  • Available in 50 durometer
  • Laser etched for lot traceability

It features ultra low extractables, excellent flexibility, longer pump life and non-yellowing formulation.

Applications include sterile fill and transfers, drug delivery, cell harvest, media processing, sanitary peristaltic pump tubing, production filtration and fermentation, bioreactor process lines.

Zertifizierungen: Meets USP Class VI and EP 3.1.9 silicone elastomer. Manufactured in an FDA registered and ISO criteria.

Bestellinformation: Available in three different lengths. Durometer 65 tubing only available on request.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge