Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Sani-Tech® Ultra-C Tubings

Lieferant: SAINT GOBAIN

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

ULT-C-1000-5 ULTRA-C-750-1
GESSULT-C-1000-5EA 0 EUR InStock
GESSULT-C-1000-5 GESSULTRA-C-750-1
Sani-Tech® Ultra-C Tubings
Schläuche Transferschlauch
Sani-Tech® Ultra-C and Ultra-65 are ultra-pure, biopharmaceutical grade silicone tubing with long pump life capabilities. Available in 50 or 65 Shore A hardness options, they are manufactured and packaged in a certified Class 7 clean room. Ultra-low TOCs ensure that contamination by extractables is kept to an absolute minimum.

  • Platinum-cured tubing
  • Ultra-low extractables
  • Excellent flexibility
  • Laser etched for lot traceability
  • Available in custom lengths and durometers
  • Superior pump life
  • Fully autoclavable and sterilisable
  • Non-yellowing formulation
  • Imparts no taste or odours
  • Temperature range from (−62 to +260 °C) −80 to +500 °F)

Sani-Tech Ultra tubing can be combined with Sani-Tech® Sani-Link® Ultra platinum-cured manifolds to create custom configurations for demanding biopharmaceutical applications. Ideal for applications such as fluid transfer in pharmaceutical, biopharmaceutical, and or laboratory settings, peristaltic pump tubing and single-use assemblies.

Hardness: 50 (shore A)

Zertifizierungen: Meets all USP Class VI, European Pharmacopoeia 3.1.9, FDA and ISO 10993 criteria.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge