Precision hotplates with separate controller
Lieferant: GESTIGKEIT HARRY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PZ35EB
GESTPZ35EBEA 2140 EUR InStock
GESTPZ35EB
Precision hotplates with separate controller
Heizgeräte Heizplatten
This precision hotplate is equipped with adjustable temperature sensors (100 to 350 °C). These sensors function as an emergency control which protects the appliance and the substance being heated from dangerous overheating.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge